Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107951.1 | NP_001101421.1 | 3 | 2585 | ||
XM_006238589.1 | XP_006238651.1 | 3 | 2584 |