Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 8 | 727 | ||
NM_015289.3 | NP_056104.2 | 7 | 694 | |||
XM_011521403.2 | XP_011519705.1 | 8 | 696 | |||
XM_011521404.2 | XP_011519706.1 | 7 | 699 | |||
XM_017022036.1 | XP_016877525.1 | 8 | 695 | |||
XM_017022037.1 | XP_016877526.1 | 7 | 699 | |||
XM_017022038.1 | XP_016877527.1 | 7 | 882 | |||
XM_017022039.1 | XP_016877528.1 | 7 | 667 | |||
GenBank mRNA | AB018313.2 | - | 7 | 645 | ||
AF280814.1 | - | 8 | 554 | |||
AF281052.1 | - | 7 | 664 | |||
BC015817.2 | - | 3 | 254 | |||
BC068559.1 | - | 7 | 663 | 5295166 | ||
BX537404.1 | - | 7 | 673 |