Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 11, 12 | 1667 | ||
NM_015291.3 | NP_056106.1 | 12, 13 | 1852 | |||
NR_109898.1 | 12, 13 | 1852 | ||||
GenBank mRNA | AB023179.2 | - | 12, 13 | 1808 | ||
AK022530.1 | - | 3, 4 | 219 | |||
AK023805.1 | - | 8, 9 | 1022 | |||
AK091848.1 | - | 12, 13 | 1810 | |||
AK292479.1 | - | 12, 13 | 1784 | |||
AL137715.1 | - | 7, 8 | 1054 | |||
AL833299.1 | - | 12, 13 | 1791 | |||
BC031991.1 | - | 4, 5 | 402 | |||
BC047363.1 | - | 12, 13 | 1852 | 5269539 | ||
BX648982.1 | - | 12, 13 | 1777 | |||
CR749259.1 | - | 11, 12 | 1612 | |||
CR749567.1 | - | 11, 12 | 1612 |