Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128184.1 | NP_001121656.1 | 2 | 409 | ||
GenBank mRNA | BC166510.1 | - | 2 | 287 | 7127430 | |
BC168710.1 | - | 2 | 390 | 8364370 |