Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 8 | 1279 | ||
NM_015291.3 | NP_056106.1 | 9 | 1464 | |||
NR_109898.1 | 9 | 1464 | ||||
GenBank mRNA | AB023179.2 | - | 9 | 1420 | ||
AK023805.1 | - | 5 | 634 | |||
AK091848.1 | - | 9 | 1422 | |||
AK292479.1 | - | 9 | 1396 | |||
AL137715.1 | - | 3 | 394 | |||
AL833299.1 | - | 9 | 1403 | |||
BC031991.1 | - | 1 | 14 | |||
BC047363.1 | - | 9 | 1464 | 5269539 | ||
BX648982.1 | - | 9 | 1389 | |||
CR749259.1 | - | 8 | 1224 | |||
CR749567.1 | - | 8 | 1224 |