Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109278.2 | NP_001102748.1 | 4 | 407 | ||
GenBank mRNA | FN806631.1 | - | 2 | 176 |