Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134933.2 | NP_001128405.1 | 3 | 168 | ||
NM_001244867.1 | NP_001231796.1 | 2 | 116 | |||
GenBank mRNA | CB729209.1 | - | 3 | 168 | ||
FQ223262.1 | - | 2 | 116 |