Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101675.1 | NP_001095145.1 | 2 | 559 | ||
GenBank mRNA | AB354637.1 | - | 2 | 559 |