Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127652.1 | NP_001121124.1 | 1066 | |||
GenBank mRNA | BC158762.1 | - | 1066 | 7105696 | ||
FQ215460.1 | - | 5, 6 | 718 | |||
FQ216242.1 | - | 5, 6 | 894 | |||
FQ216641.1 | - | 7, 8 | 1107 | |||
FQ224910.1 | - | 7, 8 | 1105 | |||
X59736.1 | - | 7, 8 | 1094 |