Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749042.3 | XP_003749090.1 | 2 | 286 | ||
XM_003749043.3 | XP_003749091.1 | 2 | 249 | |||
XM_006230934.1 | XP_006230996.1 | 2 | 362 | |||
XM_006230935.2 | XP_006230997.1 | 2 | 232 | |||
XM_006230936.1 | XP_006230998.1 | 2 | 233 | |||
XM_006230937.2 | XP_006230999.1 | 2 | 181 | |||
XM_008760196.1 | XP_008758418.1 | 1 | 670 | |||
GenBank mRNA | BC166555.1 | - | 2 | 254 | 7311621 |