Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304333.2 | NP_001291262.1 | 4 | 802 | ||
NM_015297.2 | NP_056112.1 | 4 | 798 | |||
NR_136307.1 | 4 | 802 | ||||
NR_136308.1 | 4 | 798 | ||||
XM_011517827.1 | XP_011516129.1 | 5 | 787 | |||
XM_011517828.2 | XP_011516130.1 | 4 | 891 | |||
XM_011517829.1 | XP_011516131.1 | 4 | 1049 | |||
XM_017014553.1 | XP_016870042.1 | 6 | 1081 | |||
XM_017014554.1 | XP_016870043.1 | 6 | 861 | |||
XM_017014555.1 | XP_016870044.1 | 6 | 833 | |||
XM_017014556.1 | XP_016870045.1 | 6 | 1365 | |||
GenBank mRNA | AB028968.1 | - | 4 | 669 | ||
AK294342.1 | - | 4 | 802 |