Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024160.1 | NP_077074.1 | 2 | 163 | ||
GenBank mRNA | AJ295951.1 | - | 2 | 149 | ||
U18729.1 | - | 2 | 163 |