Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019360.2 | NP_062233.2 | 3 | 255 | ||
GenBank mRNA | BC058480.1 | - | 3 | 231 | 6922382 | |
CF976717.1 | - | 3 | 259 | |||
FQ210091.1 | - | 3 | 233 | |||
FQ210263.1 | - | 3 | 232 | |||
FQ217805.1 | - | 3 | 258 | |||
FQ221395.1 | - | 3 | 234 | |||
FQ222319.1 | - | 3 | 227 | |||
FQ222901.1 | - | 3 | 276 | |||
FQ223820.1 | - | 3 | 230 | |||
K01565.1 | - | 1 | 42 | |||
M27466.1 | - | 3 | 266 | |||
M27467.1 | - | 2 | 230 |