Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109269.1 | NP_001102739.1 | 3 | 320 | ||
XM_008764170.1 | XP_008762392.1 | 3 | 335 |
Gene Symbol: LOC503116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240815.2 | XP_006240877.1 | 3 | 258 |