Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asun | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109608.1 | NP_001103078.1 | 2 | 663 | ||
XM_006237689.1 | XP_006237751.1 | 2 | 708 |