Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135174.1 | NP_001128646.1 | 3 | 299 | ||
GenBank mRNA | CB693792.1 | - | 3 | 309 | ||
CB698700.1 | - | 3 | 276 | |||
FQ221229.1 | - | 3 | 304 | |||
FQ223031.1 | - | 3 | 305 | |||
FQ223270.1 | - | 3 | 311 | |||
FQ224596.1 | - | 3 | 303 |