Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 9 | 754 | ||
NM_015303.3 | NP_056118.2 | 8 | 748 | |||
XM_005247251.3 | XP_005247308.1 | 9 | 1022 | |||
XM_005247253.4 | XP_005247310.1 | 9 | 760 | |||
XM_006713556.3 | XP_006713619.1 | 1 | 42 | |||
XM_011512601.2 | XP_011510903.1 | 1 | 42 | |||
XM_017006040.1 | XP_016861529.1 | 7 | 442 | |||
XM_017006041.1 | XP_016861530.1 | 9 | 760 | |||
GenBank mRNA | AK056661.1 | - | 9 | 726 | ||
AK292423.1 | - | 8 | 720 | |||
AL833838.1 | - | 6 | 287 | |||
BC140768.1 | - | 9 | 690 | 9021636 | ||
BX647915.1 | - | 8 | 710 |