Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134908.1 | NP_001128380.1 | 3 | 733 | ||
XM_006237192.2 | XP_006237254.1 | 4 | 1420 | |||
XM_006237193.2 | XP_006237255.1 | 4 | 1449 | |||
XM_006237194.2 | XP_006237256.1 | 4 | 1375 | |||
GenBank mRNA | BC158822.1 | - | 3 | 733 | 7376045 | |
FQ213084.1 | - | 3 | 705 |