Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107086.1 | NP_001100556.1 | 3 | 296 |
Gene Symbol: LOC100911374 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751074.3 | XP_003751122.2 | 1 | 445 | ||
XM_008768806.1 | XP_008767028.1 | 1 | 444 |