Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022504.1 | NP_071949.1 | 1 | 73 | ||
GenBank mRNA | BC058475.1 | - | 2 | 135 | 6888196 | |
FQ216992.1 | - | 1 | 75 | |||
FQ222487.1 | - | 2 | 94 | |||
X68284.1 | - | 1 | 73 |