Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135918.1 | NP_001129390.1 | 2 | 151 | ||
XM_006246034.2 | XP_006246096.1 | 2 | 295 |