Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102546354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255153.2 | XP_006255215.1 | 2 | 3317 | ||
XM_006255155.2 | XP_006255217.1 | 5 | 3014 | |||
XM_006255160.2 | XP_006255222.1 | 4 | 1266 | |||
XM_006255164.2 | XP_006255226.1 | 4 | 360 | |||
XM_008772333.1 | XP_008770555.1 | 4 | 4655 | |||
XM_008772334.1 | XP_008770556.1 | 5 | 4155 | |||
XM_008772335.1 | XP_008770557.1 | 5 | 3851 | |||
XM_008772336.1 | XP_008770558.1 | 4 | 3710 | |||
XM_008772337.1 | XP_008770559.1 | 4 | 4678 |
Gene Symbol: Zfp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100991.2 | NP_001094461.1 | 5 | 595 | ||
XM_006228733.2 | XP_006228795.1 | 2 | 3313 | |||
XM_006228735.2 | XP_006228797.1 | 4 | 4652 | |||
XM_006228737.2 | XP_006228799.1 | 4 | 3712 | |||
XM_006228738.2 | XP_006228800.1 | 5 | 3849 | |||
XM_006228740.2 | XP_006228802.1 | 4 | 1263 | |||
XM_006228743.2 | XP_006228805.1 | 4 | 360 | |||
XM_008759177.1 | XP_008757399.1 | 5 | 3011 | |||
GenBank mRNA | BC128772.1 | - | 5 | 620 | ||
BC166403.1 | - | 3 | 259 | 8363623 |