Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015306.2 | NP_056121.2 | 12 | 1618 | ||
XM_017000831.1 | XP_016856320.1 | 12 | 1623 | |||
XM_017000832.1 | XP_016856321.1 | 12 | 1620 | |||
XM_017000833.1 | XP_016856322.1 | 12 | 1115 | |||
XM_017000834.1 | XP_016856323.1 | 12 | 1115 | |||
XM_017000835.1 | XP_016856324.1 | 12 | 1626 | |||
XM_017000836.1 | XP_016856325.1 | 12 | 1626 | |||
XM_017000837.1 | XP_016856326.1 | 12 | 1626 |