Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100534.1 | NP_001094004.1 | 4 | 751 | ||
GenBank mRNA | BC159423.1 | - | 4 | 639 | ||
BC159431.1 | - | 4 | 751 | 9091211 | ||
FQ211668.1 | - | 3 | 446 | |||
FQ217699.1 | - | 3 | 446 | |||
FQ219786.1 | - | 3 | 446 | |||
FQ220709.1 | - | 3 | 446 | |||
FQ220767.1 | - | 3 | 446 | |||
FQ221206.1 | - | 3 | 445 | |||
FQ221432.1 | - | 3 | 446 | |||
FQ221889.1 | - | 3 | 446 | |||
FQ222645.1 | - | 3 | 446 | |||
FQ222683.1 | - | 3 | 446 | |||
FQ223086.1 | - | 3 | 446 | |||
FQ223813.1 | - | 3 | 446 | |||
FQ224905.1 | - | 3 | 446 | |||
FQ225844.1 | - | 3 | 446 | |||
FQ226679.1 | - | 3 | 446 | |||
FQ228074.1 | - | 3 | 446 | |||
FQ228176.1 | - | 3 | 446 | |||
FQ229585.1 | - | 3 | 446 | |||
FQ229680.1 | - | 3 | 446 | |||
FQ229824.1 | - | 3 | 446 | |||
FQ230037.1 | - | 3 | 446 | |||
FQ231936.1 | - | 3 | 446 | |||
FQ232074.1 | - | 3 | 446 | |||
FQ232489.1 | - | 3 | 446 | |||
FQ232741.1 | - | 3 | 446 | |||
FQ234236.1 | - | 3 | 446 | |||
FQ235314.1 | - | 3 | 446 |