Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318339.1 | NP_001305268.1 | 8 | 1608 | ||
NM_015308.3 | NP_056123.2 | 8 | 1602 | |||
GenBank mRNA | AB023231.2 | - | 8 | 1601 | ||
AK022987.1 | - | 1 | 46 | |||
AK054636.1 | - | 8 | 1425 | |||
AK123898.1 | - | 7 | 1269 | |||
AL137480.1 | - | 8 | 1401 | |||
BC037404.1 | - | 8 | 1433 | 5140963 |