Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012659.1 | NP_036791.1 | 2 | 411 | ||
GenBank mRNA | J00788.1 | - | 2 | 242 | ||
M25890.1 | - | 2 | 411 | |||
V01271.1 | - | 2 | 411 |