Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126289.1 | NP_001119761.1 | 12 | 1820 | ||
GenBank mRNA | BC160823.1 | - | 12 | 1820 | 7380256 |