Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013224.1 | NP_037356.1 | 4 | 374 | ||
GenBank mRNA | BC061561.1 | - | 4 | 378 | 6921494 | |
FQ210888.1 | - | 4 | 374 | |||
FQ211456.1 | - | 4 | 373 | |||
FQ216976.1 | - | 4 | 374 | |||
FQ217039.1 | - | 4 | 374 | |||
FQ217630.1 | - | 4 | 374 | |||
FQ220682.1 | - | 4 | 374 | |||
FQ220803.1 | - | 4 | 445 | |||
FQ221742.1 | - | 4 | 374 | |||
FQ222007.1 | - | 4 | 374 | |||
FQ222170.1 | - | 4 | 374 | |||
FQ222423.1 | - | 4 | 375 | |||
FQ222638.1 | - | 4 | 374 | |||
FQ223096.1 | - | 4 | 373 | |||
FQ223202.1 | - | 4 | 374 | |||
FQ223253.1 | - | 4 | 370 | |||
FQ223633.1 | - | 4 | 374 | |||
FQ224810.1 | - | 4 | 374 | |||
FQ229027.1 | - | 4 | 374 | |||
FQ233862.1 | - | 4 | 374 | |||
FQ233965.1 | - | 3 | 374 | |||
FQ234974.1 | - | 4 | 375 | |||
FQ235169.1 | - | 4 | 375 | |||
FQ235253.1 | - | 4 | 374 | |||
X02414.1 | - | 4 | 374 |
Gene Symbol: LOC100360508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728001.3 | XP_002728047.2 | 1 | 408 |
Gene Symbol: LOC100361854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730210.4 | XP_002730256.1 | 1 | 368 |
Gene Symbol: RGD1559808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_236845.5 | XP_236845.1 | 1 | 351 |
Gene Symbol: RGD1562415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_221359.6 | XP_221359.1 | 1 | 420 |