Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134882.1 | NP_001128354.1 | 3 | 359 | ||
XM_006254276.2 | XP_006254338.1 | 3 | 476 | |||
XM_006254278.2 | XP_006254340.1 | 3 | 279 | |||
XM_008771888.1 | XP_008770110.1 | 3 | 262 |