Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012963.2 | NP_037095.1 | 1192 | |||
GenBank mRNA | AF275734.1 | - | 1223 | |||
BC061779.1 | - | 1175 | ||||
BC081839.1 | - | 1192 | ||||
BC088402.1 | - | 1269 |
Gene Symbol: Hmg1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109373.1 | NP_001102843.1 | 1 | 1239 | ||
GenBank mRNA | BC168143.1 | - | 1 | 1272 | 9100911 |