Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198665.1 | NP_001185594.1 | 16, 17 | 1730 | ||
NM_001301084.1 | NP_001288013.1 | 17, 18 | 1613 | |||
NM_015313.2 | NP_056128.1 | 17, 18 | 1787 | |||
XM_006718805.3 | XP_006718868.1 | 16, 17 | 2071 | |||
XM_011542720.2 | XP_011541022.1 | 17, 18 | 1728 | |||
XM_017017420.1 | XP_016872909.1 | 15, 16 | 2243 | |||
XM_017017421.1 | XP_016872910.1 | 17, 18 | 2003 | |||
XM_017017422.1 | XP_016872911.1 | 16, 17 | 1495 | |||
GenBank mRNA | AB002380.2 | - | 12, 13 | 1149 | ||
AF180681.1 | - | 17, 18 | 1459 | |||
AK294803.1 | - | 17, 18 | 1588 | |||
AK304316.1 | - | 17, 18 | 2011 | |||
BC063117.1 | - | 16, 17 | 1733 | |||
BX647785.1 | - | 17, 18 | 1589 | |||
BX648515.1 | - | 17, 18 | 1594 |