Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000414.1 | NP_001000414.1 | 1 | 769 |
Gene Symbol: LOC103692345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763692.1 | XP_008761914.1 | 1 | 607 |
Gene Symbol: Olr836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000406.1 | NP_001000406.1 | 1 | 769 |
Gene Symbol: Olr837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000897.1 | NP_001000897.1 | 1 | 769 | ||
GenBank mRNA | AF091565.1 | - | 1 | 577 |
Gene Symbol: Olr838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000581.1 | NP_001000581.1 | 1 | 766 |