Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033075.1 | NP_001028247.1 | 1, 2 | 179 | ||
GenBank mRNA | AY623751.1 | - | 1, 2 | 179 |
Gene Symbol: Defa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033078.1 | NP_001028250.1 | 1, 2 | 164 | ||
XM_006253369.1 | XP_006253431.1 | 2, 3 | 262 | |||
GenBank mRNA | AY623755.1 | - | 1, 2 | 164 |