Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135809.1 | NP_001129281.1 | 6 | 622 | ||
GenBank mRNA | DQ537495.1 | - | 6 | 622 |
Gene Symbol: LOC100359989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233654.2 | XP_006233716.2 | 6 | 571 |