Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134726.1 | NP_001128198.1 | 1045 | |||
GenBank mRNA | BC167072.1 | - | 1045 | 9028223 |
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255656.2 | XP_006255718.1 | 4 | 886 | ||
XM_006255657.2 | XP_006255719.1 | 4 | 1644 | |||
XM_006255660.2 | XP_006255722.1 | 4 | 1610 | |||
XM_006255662.2 | XP_006255724.1 | 4 | 837 | |||
XM_006255663.2 | XP_006255725.1 | 3 | 1541 | |||
XM_008772583.1 | XP_008770805.1 | 5 | 1841 | |||
XM_008772584.1 | XP_008770806.1 | 5 | 1838 | |||
XM_008772585.1 | XP_008770807.1 | 5 | 1790 | |||
XM_008772586.1 | XP_008770808.1 | 4 | 1214 | |||
XM_574241.6 | XP_574241.4 | 4 | 853 |
Gene Symbol: Zfp879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134600.1 | NP_001128072.1 | 5 | 1379 |
Gene Symbol: Zfp952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729770.4 | XP_002729816.1 | 4 | 3219 | ||
XM_008765188.1 | XP_008763410.1 | 5 | 1361 | |||
GenBank mRNA | FQ222140.1 | - | 4 | 1191 |