Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC298116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003409.1 | NP_001003409.1 | 3 | 301 | ||
GenBank mRNA | M26836.1 | - | 3 | 301 |
Gene Symbol: LOC102548083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763747.1 | XP_008761969.1 | 365 |
Gene Symbol: LOC259244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147212.1 | NP_671745.1 | 337 | |||
GenBank mRNA | AB039824.1 | - | 337 |
Gene Symbol: LOC259246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147214.2 | NP_671747.1 | 3 | 344 | ||
GenBank mRNA | AB039822.1 | - | 3 | 337 | ||
BC088109.1 | - | 3 | 339 | 7322607 | ||
BC098654.1 | - | 3 | 344 | 7370970 | ||
BC105816.1 | - | 3 | 341 | 7367233 | ||
J00737.1 | - | 3 | 286 | |||
M26835.1 | - | 3 | 316 | |||
M26837.1 | - | 3 | 312 | |||
M26838.1 | - | 3 | 309 | |||
U31287.1 | - | 3 | 351 |
Gene Symbol: LOC500473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238221.2 | XP_006238283.1 | 412 |
Gene Symbol: Obp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033958.2 | NP_001029130.2 | 356 | |||
NM_001033959.2 | NP_001029131.1 | 356 | ||||
NM_147215.2 | NP_671748.2 | 356 | ||||
GenBank mRNA | AB039825.1 | - | 337 | |||
BC086942.1 | - | 326 | 7369206 | |||
J00738.1 | - | 356 | ||||
X14552.1 | - | 352 |
Gene Symbol: RGD1566134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203512.2 | NP_988843.2 | 337 | |||
GenBank mRNA | AB039823.1 | - | 337 | |||
BC086943.1 | - | 349 | 7372252 |