Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079943.1 | NP_001073412.1 | 8 | 1405 | ||
XM_006242666.1 | XP_006242728.1 | 7 | 1412 | |||
XM_006242667.2 | XP_006242729.1 | 7 | 3500 | |||
XM_006242668.2 | XP_006242730.1 | 8 | 2806 | |||
XM_006242670.1 | XP_006242732.1 | 8 | 1495 | |||
XM_006242671.2 | XP_006242733.1 | 6 | 1281 | |||
XM_006242672.2 | XP_006242734.1 | 7 | 3008 | |||
XM_006242673.2 | XP_006242735.1 | 6 | 2861 | |||
XM_008765981.1 | XP_008764203.1 | 7 | 3011 | |||
XM_008765982.1 | XP_008764204.1 | 7 | 1348 | |||
XM_008765983.1 | XP_008764205.1 | 7 | 2656 | |||
XM_008765984.1 | XP_008764206.1 | 7 | 2612 | |||
GenBank mRNA | BC129125.1 | - | 8 | 1405 | 7133611 |