Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134993.1 | NP_001128465.1 | 6 | 689 | ||
GenBank mRNA | BC169052.1 | - | 6 | 653 | 7386405 | |
FQ220871.1 | - | 6 | 651 |