Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134796.1 | NP_001128268.1 | 11 | 1253 | ||
XM_006252943.2 | XP_006253005.1 | 12 | 1714 | |||
XM_006252944.2 | XP_006253006.1 | 12 | 5245 | |||
XM_006252945.2 | XP_006253007.1 | 13 | 1152 | |||
XM_006252946.2 | XP_006253008.1 | 11 | 5221 | |||
XM_006252947.2 | XP_006253009.1 | 12 | 1131 | |||
XM_008771150.1 | XP_008769372.1 | 13 | 1735 | |||
XM_008771151.1 | XP_008769373.1 | 13 | 1732 | |||
XM_008771152.1 | XP_008769374.1 | 13 | 1720 | |||
XM_008771153.1 | XP_008769375.1 | 12 | 1711 | |||
XM_008771154.1 | XP_008769376.1 | 11 | 5221 |