Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134735.1 | NP_001128207.1 | 744 | |||
GenBank mRNA | BC168203.1 | - | 744 | 7115651 |