Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053861.1 | NP_446313.1 | 24 | 5927 | ||
XM_008763754.1 | XP_008761976.1 | 27 | 6809 | |||
XM_008763755.1 | XP_008761977.1 | 26 | 6536 | |||
XM_008763756.1 | XP_008761978.1 | 26 | 6532 | |||
XM_008763757.1 | XP_008761979.1 | 25 | 6258 | |||
XM_008763758.1 | XP_008761980.1 | 21 | 5164 | |||
GenBank mRNA | EU596506.1 | - | 24 | 5927 |