Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053861.1 | NP_446313.1 | 6 | 2530 | ||
XM_008763754.1 | XP_008761976.1 | 7 | 2863 | |||
XM_008763755.1 | XP_008761977.1 | 7 | 2863 | |||
XM_008763756.1 | XP_008761978.1 | 7 | 2862 | |||
XM_008763757.1 | XP_008761979.1 | 7 | 2861 | |||
XM_008763758.1 | XP_008761980.1 | 7 | 2859 | |||
GenBank mRNA | EU596506.1 | - | 6 | 2530 |