Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053861.1 | NP_446313.1 | 14 | 4528 | ||
XM_008763754.1 | XP_008761976.1 | 17 | 5410 | |||
XM_008763755.1 | XP_008761977.1 | 16 | 5137 | |||
XM_008763756.1 | XP_008761978.1 | 16 | 5133 | |||
XM_008763757.1 | XP_008761979.1 | 15 | 4859 | |||
XM_008763758.1 | XP_008761980.1 | 11 | 3765 | |||
GenBank mRNA | EU596506.1 | - | 14 | 4528 | ||
U09361.1 | - | 6 | 1321 | |||
U09401.1 | - | 2 | 229 | |||
U15550.1 | - | 3 | 731 |