Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134970.1 | NP_001128442.1 | 28 | 3801 | ||
XM_008759797.1 | XP_008758019.1 | 30 | 4113 | |||
XM_008759798.1 | XP_008758020.1 | 30 | 4110 | |||
XM_008759800.1 | XP_008758022.1 | 29 | 4017 | |||
XM_008759801.1 | XP_008758023.1 | 29 | 4014 | |||
XM_008759802.1 | XP_008758024.1 | 29 | 3975 | |||
XM_008759803.1 | XP_008758025.1 | 29 | 3893 | |||
GenBank mRNA | BC168698.1 | - | 28 | 3801 | 7938988 |