Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134981.1 | NP_001128453.1 | 16 | 2752 | ||
XM_008766280.1 | XP_008764502.1 | 15 | 2559 | |||
XM_008766281.1 | XP_008764503.1 | 14 | 2220 | |||
XM_008766283.1 | XP_008764505.1 | 14 | 2784 | |||
XM_008766284.1 | XP_008764506.1 | 15 | 2351 | |||
GenBank mRNA | BC168748.1 | - | 15 | 2351 | 6921198 |