Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100125368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139487.1 | NP_001132959.1 | 4 | 802 | ||
XM_006240835.2 | XP_006240897.1 | 4 | 976 | |||
XM_008765080.1 | XP_008763302.1 | 4 | 1235 | |||
GenBank mRNA | BC128765.1 | - | 4 | 802 | 7123097 |
Gene Symbol: LOC102551539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765145.1 | XP_008763367.1 | 700 | |||
XM_008765146.1 | XP_008763368.1 | 1235 |