Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-O1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008856.1 | NP_001008856.1 | 2 | 166 | ||
XM_006255910.1 | XP_006255972.1 | 1 | 118 |
Gene Symbol: RT1-S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008857.1 | NP_001008857.1 | 2 | 157 | ||
XM_008772705.1 | XP_008770927.1 | 2 | 2752 |