Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134596.1 | NP_001128068.1 | 1, 2 | 410 | ||
XM_006244471.2 | XP_006244533.1 | 1, 2 | 405 |