Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100136.2 | NP_001093606.2 | 5 | 482 | ||
GenBank mRNA | AA933157.1 | - | 4 | 416 | ||
BE098288.1 | - | 4 | 416 | |||
FQ211057.1 | - | 5 | 437 | |||
FQ220566.1 | - | 5 | 503 |