Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC690460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109593.1 | NP_001103063.1 | 1 | 385 |
Gene Symbol: LOC103694398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772850.1 | XP_008771072.1 | 1 | 355 | ||
XM_008772851.1 | XP_008771073.1 | 1 | 355 |
Gene Symbol: LOC686720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772839.1 | XP_008771061.1 | 1 | 466 | ||
XM_008772840.1 | XP_008771062.1 | 2 | 391 |
Gene Symbol: LOC687631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772838.1 | XP_008771060.1 | 1 | 410 |
Gene Symbol: LOC689927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109555.1 | NP_001103025.1 | 466 |
Gene Symbol: LOC690415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109590.1 | NP_001103060.1 | 1 | 328 | ||
XM_008772833.1 | XP_008771055.1 | 1 | 328 |
Gene Symbol: LOC690431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256270.2 | XP_006256332.1 | 1 | 355 | ||
XM_008772842.1 | XP_008771064.1 | 2 | 292 | |||
XM_008772843.1 | XP_008771065.1 | 2 | 280 | |||
XM_008772844.1 | XP_008771066.1 | 1 | 355 | |||
XM_008772845.1 | XP_008771067.1 | 2 | 259 | |||
XM_008772846.1 | XP_008771068.1 | 1 | 355 | |||
XM_008772847.1 | XP_008771069.1 | 2 | 259 |
Gene Symbol: LOC690478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109595.1 | NP_001103065.1 | 1 | 400 |